Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
Order Info
Packaged in bags of ten
Certifications
Certified DNase- and RNase-free
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 384 |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| Well Shape | Conical Bottom (V-well) |
| For Use With (Application) | For high throughput applications requiring added volume |
| Show More |