Learn More
Corning™ Trace Elements
BIOPHARMA
Product Code. 15353641
Change view
Click to view available options
Description:
Trace Elements A
Trace Elements B
Trace Elements C
Unit Size:
100mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15353641 Trace Elements C 100mL
15333641 Trace Elements A 100mL
15343641 Trace Elements B 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15353641 Supplier Corning™ Supplier No. 25023CI
  • ONLINE EXCLUSIVE PRICE
    802.00 NOK
    369.17 NOK / 100mL
    Save 432.83 NOK
    53% Off
 Restricted Item
In Stock
For Research applications
cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
  • Trace Elements A includes Cupric Sulfate, Ferric Citrate, Sodium Selenite, and Zinc Sulfate
  • Trace Elements B includes Ammonium Molybdate, Ammonium Vanadate, Manganese Sulfate, Nickel Sulfate, Sodium Silicate, Stannous Chloride and Hydrochloric Acid
  • Trace Elements C includes Aluminum Chloride, Barium Acetate, Cadmium Chloride, Chromic Chloride, Cobalt Dichloride, Germanium Dioxide, Potassium Bromide, Potassium Iodide, Rubidium Chloride, Silver Nitrate, Sodium Fluoride, and Zirconyl Chloride
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C
Description Trace Elements C
Form Liquid
Product Type Trace Element
Sterility Sterile-filtered
Format Plastic Bottle
Quantity 100 mL