Learn More
Corning™ CellBIND™ Surface Microplates
BIOPHARMAGREENER_CHOICE
Product Code. 10234832 Shop All Corning Products
Change view
Click to view available options
Culture Area:
0.95 cm2
1.9 cm2
3.8 cm2
9.5 cm2
Capacity (Metric):
0.28 mL (Well)
0.57 mL (Well)
2.9 mL (Well)
6.9 mL
Volume (Metric) Working:
0.19 to 0.28 mL
0.38 to 0.57 mL
0.76 to 1.14 mL
1.9 to 2.9 mL
Well Count:
6
12
24
48
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area Capacity (Metric) Volume (Metric) Working Well Count unitSize
10234832 9.5 cm2 2.9 mL (Well) 1.9 to 2.9 mL 6 Pack of 50
10739864 3.8 cm2 6.9 mL 0.76 to 1.14 mL 12 Pack of 50
10224882 1.9 cm2 0.57 mL (Well) 0.38 to 0.57 mL 24 Pack of 50
10251443 0.95 cm2 0.28 mL (Well) 0.19 to 0.28 mL 48 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10234832 Supplier Corning™ Supplier No. 3335
Only null left
Add to basket
Edge
Add to basket
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Certifications
Certified nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 2.9 mL (Well)
Color Clear
Culture Area 9.5 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment CellBIND
Type Multi-well Cell Culture Plate
Volume (Metric) Working 1.9 to 2.9 mL
Well Count 6
Includes Lid
No. per Case 50/Cs.
No. per Pack 5/Pk.