Learn More
Corning™ Cell Expansion Bags
BIOPHARMA
Product Code. 17243584 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1 L
3 L
5 L
Fill Volume:
190 mL
381 mL
633 mL
1252 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Fill Volume Unit Size
17243584 1 L 381 mL Each
17253584 3 L 633 mL Each
17233584 500 mL 190 mL Each
17263584 5 L 1252 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17243584 Supplier Corning™ Supplier No. 9120085
  • ONLINE EXCLUSIVE PRICE
    1420.00 NOK
    1348.53 NOK / Pack of 1
    Save 71.47 NOK
    5% Off
Estimated Shipment: 08-02-2027
Features a single-web polyolefin film with very good clarity when filled for viewing cultures, suited for scalable expansion
Cell expansion bags are intended for the expansion and culture of non-adherent cells. The bags are made from single-web polyolefin, gas permeable film. The integrated tubing allows for functionally closed system filling, feeding, and sampling.
Features and Benefits
  • Cell expansion observed with multiple cell models
  • Gas permeable film
  • Reusable sampling valve for in-process testing
  • Tubing for sterile weld connections
  • Scalable, user friendly
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20° to 40°C
Certifications/Compliance ANSI, AMI, ISO 11137
For Use With (Application) Expansion and culture of non-adherent cells, ideal for basic research biopharmaceutical applications
Includes Reusable sampling valve
Tubing Style Integrated
Fill Volume 381 mL
Film Description Single web polyolefin gas permeable film with good clarity
No. per Case 1/Ea.
No. per Pack 1/Pk.
Port Detail Port 1- two leads with female luers and small pinch clamps, port 2 -dockable fill line with sampling valve and small pinch clamp
Quantity Each
Sterilization Method Radiation
Product Type Cell Expansion Bag
Capacity (Metric) 1 L