Learn More
Corning™ Calcein AM Fluorescent Dye
Product Code. 10520286 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
Product Code. Quantity unitSize
10520286 500 μg 500µg
10530286 1 mg 1mg
2 options
Product Code. 10520286
Brand: Corning™ 354216
Reliable and consistent dye used for the detection of live cells and key functional activities in variety of cell-based assays
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
For Use With (Equipment) Falcon FluoroBlok™ Cell culture inserts and insert systems to label cells
Quantity 500 μg
For research use only.