Learn More
Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners
Product Code. 10272703 Shop All Corning Products
Change view
Click to view available options
Closure Size:
13-415
18-415
24-400
24-410
Height (Metric):
12.5 mm
13.5 mm
17.5 mm
18.5 mm
Unit Size:
Pack of 12
Pack of 144
Pack of 192
Pack of 288
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Height (Metric) Unit Size
10272703 13-415 13.5 mm Pack of 288
10795943 24-400 12.5 mm Pack of 12
15124606 24-410 18.5 mm Pack of 144
11931899 18-415 17.5 mm Pack of 192
11333865 18-415 17.5 mm Pack of 192
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10272703 Supplier Corning™ Supplier No. 999813
  • ONLINE EXCLUSIVE PRICE
    3615.00 NOK
    3376.90 NOK / Pack of 288
    Save 238.10 NOK
    6% Off
Estimated Shipment: 19-11-2026
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.
TRUSTED_SUSTAINABILITY
Specifications
Closure Size 13-415
Liner Material PTFE
Product Type Screw Cap
Autoclavable Autoclavable
Material Phenolic
Color Black
Height (Metric) 13.5 mm
No. per Pack 288/Pk.
No. per Case 288/Cs.
Tip Material PTFE, Rubber
For Use With (Equipment) Glass Tubes