Learn More
Corning™ Black Phenolic Threaded Screw Caps
GREENER_CHOICE
Product Code. 17213604 Shop All Corning Products
Change view
Click to view available options
Neck Finish:
15-415
18-415
28-410
38-430
40-400
No. per Pack:
12/Pk.
288/Pk.
32/Pk.
72/Pk.
Unit Size:
Pack of 12
Pack of 192
Pack of 288
Pack of 72
Product Code. Neck Finish No. per Pack unitSize
17213604 38-430 12/Pk. Pack of 72
17293594 28-410 12/Pk. Pack of 12
17223604 40-400 72/Pk. Pack of 72
17263594 18-415 32/Pk. Pack of 192
10632044 15-415 288/Pk. Pack of 288
5 options
Product Code. 17213604
Brand: Corning™ 9999381M
Only null left
Add to basket
Add to basket
Resist effects of temperature and steam in autoclaving
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Phenolic Resin
No. per Pack 12/Pk.
Liner Material Rubber
Neck Finish 38-430
Height (Metric) 27 mm
Product Type Screw Cap