Learn More
Corning™ Black 384-Well Polypropylene Assay Plate
Product Code. p-2360197 Shop All Corning Products
Change view
Click to view available options
Lid:
With Lid
Without Lid
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 50
Product Code. Lid Sterility unitSize
10400672 Without Lid Non-sterile Pack of 100
11363725 With Lid Sterile Pack of 50
2 options
Product Code. 10400672
Brand: Corning™ 3658
Only null left
Add to basket
Add to basket
Lower compound sticking and higher solvent resistance than polystyrene plates
  • Ideal for fluorescence assays and for compound storage
  • Round-bottom wells
  • Untreated
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Color Black
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 0.015 to 0.07 mL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round
For Use With (Application) For fluorescence assays and compound storage
Quantity 100/Cs.