Description
- Ideal for fluorescence assays and for compound storage
- Round-bottom wells
- Untreated
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 16 x 24 |
| Color | Black |
| Material | Polypropylene |
| No. of Wells | 384 |
| Sterility | Sterile |
| Surface Treatment | CellBIND |
| Well Shape | Flat Bottom (F-well) |
| For Use With (Application) | For fluorescence assays and compound storage |