Learn More
Corning™ BioCoat™ Poly-D-Lysine/Laminin Culture Slide
Description
Corning™ BioCoatTM 8-well culture slide with a uniform application of PDL/Laminin. Glass slide with polystyrene vessel, lid, and safety removal tool. 4/pack, 12/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Shelf life: >= 3 months from date of shipment
Specifications
Specifications
| Type | Culture Slide |
| For Use With (Application) | Adhesion, growth and differentiation of neuronal and transfected cell lines |
| Storage Requirements | 2° to 8°C |
| No. per Pack | 3/Pk. |
| No. per Case | 12/Cs. |
| No. of Wells | 8 |