Learn More
Corning™ BioCoat™ Control Insert, PET Membrane
GREENER_CHOICE
Product Code. 10303042
Change view
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
8 μm
No. of Wells:
6
24
Inserts per Plate:
6
12
Quantity:
2
4
Unit Size:
Pack of 24
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size No. of Wells Inserts per Plate Quantity unitSize
10303042 0.4 μm 24 12 2 Pack of 24
10393032 0.4 μm 6 6 4 Pack of 24
10109361 1 μm 6 6 4 Pack of 24
10406421 1 μm 24 12 2 Pack of 24
10047911 3 μm 6 6 4 Pack of 24
10057911 3 μm 24 12 2 Pack of 24
10574271 8 μm 6 6 4 Pack of 24
10274901 8 μm 24 12 2 Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10303042 Supplier Corning™ Supplier No. 354572
Only null left
Add to basket
Add to basket
Use alongside BD BioCoat™ inserts when studying effects of the ECM component present on the BD BioCoat cell culture insert
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel™ matrix, Fibronectin, Collagen, and Laminin.
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
TRUSTED_SUSTAINABILITY
Specifications
Inserts per Plate 12
Culture Area 0.3 cm2
Material PET
Pore Size 0.4 μm
Type Well Plate
Quantity 2
Storage Requirements 2° to 8°C
No. per Pack 12/Pk.
No. per Case 24/Cs.
Surface Coating BioCoat Control
No. of Wells 24