Learn More
Corning™ Basic Fibroblast Growth Factors (bFGF), Human Recombinant
Product Code. 11513620 Shop All Corning Products
Change view
Click to view available options
Quantity:
10 μg
10 x 10 μg
5 x 10 μg
Unit Size:
100µg
10µg
50µg
Product Code. Quantity unitSize
11513620 10 x 10 μg 100µg
11563530 10 μg 10µg
11503620 5 x 10 μg 50µg
3 options
Product Code. 11513620
Brand: Corning™ 356061
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Isolated from pituitary glands
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Growth factors are proteins that bind to receptors found on the surface of non-hematopoietic cells. Each family of growth factors affect specific cell types, e.g., nerve growth factors (NGF) affect nerve cell types, and epidermal growth factors (EGF) affect epithelial cell types.
Corning offers a range of growth factors that are derived from different species, both natural and recombinant. These growth factors have been used to promote long-term proliferation in cell culture and to induce cell differentiation in a variety of cell types under serum-free or reduced serum conditions.
Order Info
  • FGF is supplied lyophilized, bFGF in 3.6mM Tris, PBS pH 7 buffer
  • bFGF (human) is lyophilized
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Product Type Growth Factor
Formulation Lyophilized from phosphate buffered saline, carrier free, reconstitute in dH2O or serum-free medium
For Use With (Application) Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
Purity or Quality Grade >95% by SDS-PAGE
Quantity 10 x 10 μg
Reactivity Human
Source E. coli