Learn More
Corning™ Bar Coded Storage Tubes: 1D/2D Bar Coded
For enhanced sample storage, identification and retrieval
12499.40 NOK
| Product Code | Brand | Description | Packaging Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Description | Packaging Quantity | Price | Quantity & Availability | ||||
|
11774945
|
Corning™
8502 |
Bar Coded Storage Tubes, w/ Screw caps | 96/Pk., 10Pk./Cs. |
14797.07 NOK 12499.40 NOK / Pack of 960 Save 2297.67 NOK 15% Off |
Estimated Shipment: 02-11-2026
Log in to see stock. |
||||
Description
Feature a synchronized coding scheme that minimizes confusion and the risk of duplication, while enabling safe and secure sample identification.
- Maximum Identification — Synchronized 2D and linear bar code
- Maximum Information — 14 x 14 dot 2D bar code
- Bar Code Stability — Laser-etched, not an attached label
- Compound Compatibility — Polypropylene construction throughout creates inert vessel
- Temp. tolerance: -180 to +110°C (screw cap); -80 to +110°C (TPE cap)
- Length: 43.9mm
- 96/rack
Specifications
| Polypropylene | |
| Non-sterile |