Description
- Tubes constructed from polypropylene or polystyrene
Specifications
Specifications
| Color | Clear |
| Capacity (Metric) | 5 mL |
| Length (Metric) | 75 mm |
| Material | Polystyrene |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Diameter (Metric) | 12 mm |
| Product Type | Test Tube |