Learn More
Corning™ Adapters for LSE™ Compact Centrifuges
Product Code. 11317691 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
0.2 mL tubes
0.5/0.4 mL tubes
10/15 mL tubes in 50 mL rotor
5/7/10 mL tubes in 15 mL rotor
Unit Size:
Each
Pack of 6
Product Code. For Use With (Equipment) unitSize
11317691 5/7/10 mL tubes in 15 mL rotor Each
15292479 0.5/0.4 mL tubes Pack of 6
15202489 0.2 mL tubes Pack of 6
15703338 10/15 mL tubes in 50 mL rotor Pack of 6
4 options
Product Code. 11317691
Brand: Corning™ 480142
For use with Corning LSE high speed microcentrifuge.
The individual adaptors for tubes are used in the Corning LSE High Speed Microcentrifuge.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
For Use With (Equipment) 5/7/10 mL tubes in 15 mL rotor
No. per Pack 6/Pk.