Description
- Designed to remove Stripwell™ strips from the egg crate holder
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Product Type | Strip Ejector |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Well Shape | Flat Bottom (F-well) |
| For Use With (Application) | Designed to remove Stripwell™ strips from the egg crate holder |
| No. per Case | 5/Cs. |
| No. per Pack | 5/Pk. |
| Show More |