Learn More
Corning™ 96-Well Nonbinding Surface (NBS™) Microplates
Product Code. 10261692 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Clear
White
Well Shape:
Flat Bottom (F-well)
Round
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Well Shape Unit Size
10261692 Clear Flat Bottom (F-well) Pack of 100
10409331 White Flat Bottom (F-well) Pack of 100
10276091 Black Flat Bottom (F-well) Pack of 100
10111822 White Round Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10261692 Supplier Corning™ Supplier No. 3641
  • ONLINE EXCLUSIVE PRICE
    9805.00 NOK
    9160.76 NOK / Pack of 100
    Save 644.24 NOK
    6% Off
View all promotions
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
Bulk Quote Request
Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Packaging 4 inner packs of 25
    Binding Type NBS
    Color Clear
    For Use With (Equipment) Plate readers, liquid handlers and robotic systems
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Temperature Range (English) 39°F to 99°F
    Temperature Range (Metric) 4° to 37°C
    Well Color Clear