Learn More
Corning™ 96-Well Nonbinding Surface (NBS™) Microplates
Shop All Corning Products
Click to view available options
Color:
Black
Clear
White
Well Shape:
Flat Bottom (F-well)
Round
Unit Size:
Pack of 100
Description
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | White |
| For Use With (Equipment) | Plate readers, liquid handlers and robotic systems |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 75 to 200 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-binding |
| Show More |