Learn More
Corning™ 96-Well Next Generation CrystalEX Protein Crystallization Microplates
Product Code. 10471963 Shop All Corning Products
Change view
Click to view available options
Well Shape:
Conical Flat
Round
Round, Flat Bottom (F-well)
Volume (Metric) Well:
1 μL
2 μL
4 μL
Material:
Cycloolefin Copolymer
PZero
Polymer
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Shape Volume (Metric) Well Material Unit Size
10471963 Conical Flat 1 μL Polymer Pack of 50
10057412 Round, Flat Bottom (F-well) 2 μL PZero Pack of 50
10436593 Conical Flat 2 μL PZero Pack of 50
10327152 Round 4 μL Cycloolefin Copolymer Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10471963 Supplier Corning™ Supplier No. 3550
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Optimized for high-throughput sitting-drop protein crystallization
  • Corning™ 96-well and 384-well crystallization microplates are optimized for high throughput protein crystal growth and screening
  • Designed for sitting drop applications
  • Meets ANSI/SBS standard footprint dimensions for microplates
  • Ideal for fully automated crystal screening
  • Multiple formats and versatility for custom options to maximize crystal formation, identification and analysis, and harvesting
  • - Choose from four unique protein well shapes
    - Options include one or three protein wells per reservoir well
  • Cyclic olefin copolymer (COC) offers strong chemical compatibility and good optical clarity
  • Reservoir numbers are embossed on each individual well for easy identification
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Lid Without Lid
Array 8 x 12
Color Clear
Material Polymer
Volume (Metric) Well 1 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Conical Flat
Includes Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
No. per Case 50/Cs.
No. per Pack 10/Pk.