Learn More
Proteintech CoraLite™488-Phalloidin (green)
Product Code. 17511176 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
300 Tests
Unit Size:
300 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17511176 300 Tests 300 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17511176 Supplier Proteintech Supplier No. PF00001300T
 Restricted Item
Estimated Shipment: 09-11-2026
Phalloidin is a toxin isolated from mushrooms that binds F-Actin in the cytoskeleton.
  • This product is conjugated to our fluorescent CoraLite(R) dye to stain F-actin for labeling, identifying and quantifying actin filaments in immunostaining studies.
  • Compared to many fluorescent dyes, CoraLite(R) dyes have advantages in brightness, light stability and water solubility.
  • Excitation/Emission maxima wavelengths are 490 nm/515 nm
Specifications
For Use With (Application) Immunofluorescence
Quantity 300 Tests
Emission 515 nm