missing translation for 'onlineSavingsMsg'
Learn More

COPS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7088001 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16846863 20 μL 20µL
16836863 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16846863

Brand: Proteintech 104641AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CSN4 is a subunit of the JUN activation-domain binding protein 1 (JAB1), containing signalosome complex (or COP9 complex) which has kinase activity and phosphorylates C-JUN. The signalosome complex may play a role in signal transduction.

Specifications

Antigen COPS4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene COPS4
Gene Accession No. O88544, Q68FS2, Q9BT78
Gene Alias COPS4, CSN4, SGN4, Signalosome subunit 4
Gene Symbols COPS4
Host Species Rabbit
Immunogen COPS4 Fusion Protein Ag0735
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26891, 360915, 51138
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.