Learn More
Copper Silicon bar, 3.8x3.8x30cm, Thermo Scientific™
Product Code. 15461927
Change view
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461927 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15461927 Supplier Thermo Scientific Alfa Aesar Supplier No. 043767.KG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Copper Silicon bar
Name Note 3.8x3.8x30cm
Avg. Mol. Wt. or Mol. Wt. Range Cu:Si; 97:3 wt%
Quantity 1 Pc.
Weight ≈3.7kg/bar
Form Bar
MDL Number MFCD02684526
Synonym Everdur™ Silicon Bronze