missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Trifluoromethanesulfonate, TRC

Product Code. 30358013
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
Product Code. Quantity unitSize
30358013 25 g 25g
30349947 5 g -
30356329 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 30358013

Brand: Toronto Research Chemicals TRCC68537525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34946-82-2
Formula Weight 360.8336
InChI Formula InChI=1S/2CHF3O3S.Cu/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2
IUPAC Name copper;trifluoromethanesulfonate
Molecular Formula C2CuF6O6S2
Molecular Weight (g/mol) 361.68
SMILES [Cu+2].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Synonym 1,1,1-Trifluoromethanesulfonic Acid Copper(2+) Salt (2:1), Trifluoromethanesulfonic Acid Copper(2+) Salt, Bis(trifluoromethanesulfonato)copper, Copper Bis(trifluoromethanesulfonate), Copper Ditriflate, Copper(2+) Bis(trifluoromethanesulfonate), Copper(2+) Triflate, Copper(2+) Trifluoromethylsulfonate, Copper(II) Triflate, Cupric Bis(trifluoromethanesulfonate), Cupric Triflate, Cupric Trifluoromethanesulfonate, Cupric Trifluoromethylsulfonate, Trifluoromethanesulfonic Acid Copper Salt (2:1), Trifluoromethanesulfonic Acid Copper(2+) Salt
Chemical Name or Material Copper(II) Trifluoromethanesulfonate

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.