Learn More
Copper(II) phthalocyanine, BLD Pharm™
Product Code. 30781255
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30781255 100 g 100g
30777179 500 g 500g
30770832 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30781255 Supplier BLD Pharm Supplier No. BD147935100g
 Restricted Item
Estimated Shipment: 25-11-2026
Dye content 98%+
Specifications
CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Quantity 100 g
SMILES C1(/C2=N/C3=N/C(C4=CC=CC=C43)=N\C5=C6C=CC=CC6=C([N-]5)/N=C(C7=CC=CC=C78)\N=C8/N=C9\[N-]2)=C9C=CC=C1.[Cu+2]
Molecular Weight (g/mol) 576.07
Chemical Name or Material Copper(II) phthalocyanine