Learn More
Copper(II) phosphate, 98%
Description
Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.
Solubility
Insoluble with water and ethanol. Soluble in ammonia, ammonium hydroxide, acetone.
Notes
Hygroscopic. Incompatible with strong bases, metals, alkali metals and powdered metals.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 2500 g |
| Linear Formula | Cu3(PO4)2 |
| Merck Index | 14,2648 |
| Solubility Information | Insoluble with water and ethanol. Soluble in ammonia,ammonium hydroxide,acetone. |
| Formula Weight | 380.58 |
| Percent Purity | 98% |
| Odor | Odorless |
| Chemical Name or Material | Copper(II) phosphate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.