missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%

Product Code. 11331859
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331859 500 g 500g
11341859 50 g 50g
11321859 2 kg 2kg
11311859 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11331859 Supplier Thermo Scientific Alfa Aesar Supplier No. 012523.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Use As a Catalyst, Nitrating Agent For Aromatic organosilicon Compounds, Nickel-Plating Baths | CAS: 19004-19-4 | CuN2O6·2.5H2O

Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black and producing a burnished effect on ironCopper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19004-19-4
Molecular Formula CuN2O6·2.5H2O
MDL Number MFCD00149671
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

Specifications

Boiling Point 170°C (decomposition)
Melting Point ∽115°C
Physical Form Crystalline Lump
Quantity 500 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water and alcoholSoluble in water,ammonia and ethanol.
Formula Weight 232.59 (187.55 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Density 2.32 g/cm3
Chemical Name or Material Copper(II) nitrate hemi(pentahydrate), trihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.