Learn More
Copper(II) Chloride Dihydrate, Supelco™
Product Code. 15874280
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874280 1 kg 1kg
15864280 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15874280 Supplier Supelco Supplier No. 1027331000
 Restricted Item
Estimated Shipment: 20-11-2026
For analysis EMSURE™ ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10125-13-0
Specifications
Melting Point 598°C
Quantity 1 kg
MDL Number MFCD00149674
Synonym Copper(II) chloride dihydrate, Copper dichloride dihydrate, Copper(II) chloride dihydrate
Molecular Weight (g/mol) 170.48
Grade Research
Density 2.53 g/cm3 at 20°C
Chemical Name or Material Copper(II) Chloride Dihydrate