Learn More
Copper(II) Acetylacetonate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 13395-16-9 |
| Melting Point | 245°C (Decomposition) |
| Boiling Point | 160°C/13 hPa |
| MDL Number | MFCD00000016 |
| Quantity | 100 g |
| Synonym | Copper(II) acetylacetonate, Bis(acetylacetonato)copper(II), Bis(2,4-pentanedionato) copper(II), Copper(II) acetylacetonate |
| Molecular Weight (g/mol) | 261.76 |
| Grade | Research |
| Chemical Name or Material | Copper(II) Acetylacetonate |