missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Chloride, ≥97.0%, Honeywell™ Fluka™

Product Code. 15690870 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15690870 1 kg Poly Bottle 1kg
15600880 250 g Poly Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15690870 Supplier Honeywell-Fluka Supplier No. 082531KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 98.996
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]

Specifications

Boiling Point 1490°C
Melting Point 430°C
Color Gray
pH 5.0 (at 50.00g/L 20°C)
Quantity 1 kg
Linear Formula CuCl
UN Number UN2802
Solubility Information Slightly soluble in water, 0.47g/L at 20°C (USP brand)
Formula Weight 99g/mol
Percent Purity ≥97.0%
Packaging Poly Bottle
Appearance Small beads
Vapor Pressure 1.3mmHg (546°C)
Density 3.530g/cu.cm
Chemical Name or Material Copper(I) Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.