missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Copper Nickel, Hiduron 191, C72420, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Copper Nickel, Hiduron 191, C72420, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Zinc/Chromium/Cobalt/Phosphorus/Carbon/Magnesium/Silicon/Iron/Aluminium/Manganese/Nickel/Hydrogen/Nitrogen/Oxygen/Tungsten/Zirconium/Antimony/Molybdenum/Niobium/Scandium/Titanium/Vanadium/Arsenic/Cadmium/Bismuth/Silver/Tin/Boron/Lead/Selenium/Copper |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zinc 0.001 %/Chromium 0.0013 %/Cobalt 0.002 %/Phosphorus 0.003 %/Carbon 0.004 %/Magnesium 0.0059 %/Silicon 0.017 %/Iron 0.96 %/Aluminium 1.6 %/Manganese 3.79 %/Nickel 14.5 %/Hydrogen (<0.0010) %/Nitrogen (<0.0010) %/Oxygen (<0.0010) %/Tungsten (<0.0010) %/Zirconium (<0.0010) %/Antimony (<0.0050) %/Molybdenum (<0.0050) %/Niobium (<0.0050) %/Scandium (<0.0050) %/Titanium (<0.0050) %/Vanadium (<0.0050) %/Arsenic (<0.0100) %/Cadmium (0.001) %/Bismuth (0.002) %/Silver (0.002) %/Tin (0.002) %/Boron (0.003) %/Lead (0.003) %/Selenium (0.003) %/Copper (79.6) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction