Learn More
Sampling Systems™ Copper Braided Cable
Product Code. p-7138898 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
25 m
50 m
Unit Size:
25m
50m
Product Code. 15869341
Brand: Sampling Systems™ 5070C25
Only null left
Add to basket
Add to basket
Sampling Systems™ Copper Braided Cable is for use with the Hand reel Ex in Ex rated environments.
  • Braided cable constructed of copper wire
  • For use with Hand Reel Ex in Ex rated environments
Specifications
Product Type Cable
Material Copper
Description Copper cable
Length (Metric) 25 m