Learn More
Sampling Systems™ Copper Braided Cable
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
25 m
50 m
Unit Size:
25m
50m
Description
- Braided cable constructed of copper wire
- For use with Hand Reel Ex in Ex rated environments
Specifications
Specifications
| Product Type | Cable |
| Material | Copper |
| Description | Copper cable |
| Length (Metric) | 25 m |