missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, High Silicon Bronze A, CDA 655, C65500, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, High Silicon Bronze A, CDA 655, C65500, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Aluminium/Nickel/Tin/Lead/Iron/Zinc/Manganese/Silicon/Copper/Vanadium/Antimony/Arsenic/Bismuth/Boron/Cadmium/Calcium/Carbon/Chromium/Cobalt/Hydrogen/Magnesium/Molybdenum/Nitrogen/Oxygen/Palladium/Phosphorus/Selenium/Silver/Sulfur/Titanium |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.002 %/Nickel 0.0034 %/Tin 0.009 %/Lead 0.018 %/Iron 0.055 %/Zinc 0.149 %/Manganese 0.98 %/Silicon 3.15 %/Copper 95.5 %/Vanadium (<50) ppm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction