Learn More
Corning™ CoolRack™ Tall Modules
BIOPHARMA
Product Code. 15572831
Change view
Click to view available options
Holds:
1 250 mL Centrifuge Tube
1 250 mL Easy Grip Storage Bottle
12 x 15 mL Centrifuge Tubes
12 x 5 mL Centrifuge Tubes, SBS-compatible
4 x 50 mL Centrifuge Tubes
9 x 15 mL Centrifuge Tubes
Design:
CoolRack 15 mL
CoolRack 250 mL
CoolRack 250 mL PF
CoolRack 50 mL
CoolRack L
CoolRack XT 5 mL
Unit Size:
Each
Product Code. Holds Design unitSize
15572831 4 x 50 mL Centrifuge Tubes CoolRack 50 mL Each
15552831 12 x 5 mL Centrifuge Tubes, SBS-compatible CoolRack XT 5 mL Each
15562831 9 x 15 mL Centrifuge Tubes CoolRack 15 mL Each
15582831 1 250 mL Centrifuge Tube CoolRack 250 mL PF Each
15592831 1 250 mL Easy Grip Storage Bottle CoolRack 250 mL Each
15532841 12 x 15 mL Centrifuge Tubes CoolRack L Each
6 options
Product Code. 15572831
Brand: Corning™ 432062
Only null left
Add to basket
Add to basket
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Corning CoolRack thermo-conductive centrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
  • Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible
TRUSTED_SUSTAINABILITY
Specifications
Holds 4 x 50 mL Centrifuge Tubes
Quantity 1/cs.
Design CoolRack 50 mL
Type Tall Module