Learn More
Corning™ CoolRack™ CF Cryogenic Vial and FACS Tube Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Brand:  Corning™ 432049
Additional Details : Weight : 0.47340kg
 View more versions of this product
Product Code. 15542821
  • 1990.17 NOK / Each
Estimated Shipment: 28-10-2024
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
  • Ensure +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Specifications
    Specifications
    Capacity: 15 cryogenic vials or FACS tubes
    CF15
    Cryogenic Vial, FACS Tube Module