Learn More
Corning™ Extension Collar for CoolBox™ XT System
Description
For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.
Specifications
Specifications
| Color | Orange |
| Type | Extension Collar |
| For Use With (Application) | Extend the height when using tall tube modules |
| For Use With (Equipment) | CoolBox XT System |
| Quantity | 1/Pk. |
| Dimensions (L x W x H) | 20.0 x 16.0 x 7.0 cm (7.8 x 6.2 x 2.7 in.) Interior: 14.0 x 9.4 x 7.0 cm (5.5 x 3.7 x 2.7 in.) |