Learn More
SymLine Continuous Level Control Accessories
Click to view available options
Includes:
1 x banana plug, 1 x clamp
1 x clamp, 1 ring ID 10 mm
1 x clamp, 1 x button plug
1 x clamp, 1 x ring ID 5 mm
2 x clamps
2x 10 mm press button
ATEX zone 0, ATEX Ex II 1 G Ex ia IIC T1-T6 Ga
ATEX, II 2 G EEx ia IIC T1-T6, NAMUR
Fitting for continuous level control
Ground cable
Grounding cable and clamp
Product Type:
Antistatic Floor Mat
Capacitive Disc Sensor
Capacitive Rod Sensor
Earthing Cable
Grounding Cable
Grounding Plug
Grounding Strap
Safety Waste Cap
Unit Size:
Each
Description
Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!
Specifications
Specifications
| Description | Grounding cable, 1 x clamp, 1 ring ID 10 mm, L = 1,5 m |
| Length (Metric) | 1.5 m |
| Includes | 1 x clamp, 1 ring ID 10 mm |
| For Use With (Equipment) | Safety Waste Caps and Connection Rings Funnels |
| Product Type | Grounding Cable |
Product Content Correction
Product Title
SymLine Continuous Level Control Accessories >