Learn More
Contec™ QuickConnect™ Mop Head Frames
Product Code. 17535635 Shop All Contec Products
Change view
Click to view available options
Material:
Stainless Steel
Stainless Steel (304)
For Use With (Equipment):
All Styles of Edgeless Mop Heads
Medium VertiKlean, Medium or Large VertiKlean MAX
Regular VertiKlean MAX Head Frame
Length (English):
7 in.
7.5 in.
11.5 in.
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material For Use With (Equipment) Length (English) Unit Size
17535635 Stainless Steel Regular VertiKlean MAX Head Frame 7 in. Each
16615504 Stainless Steel Regular VertiKlean MAX Head Frame 7.5 in. Each
16671123 Stainless Steel (304) All Styles of Edgeless Mop Heads 7 in. Each
16030852 Stainless Steel Medium VertiKlean, Medium or Large VertiKlean MAX 11.5 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17535635 Supplier Contec™ Supplier No. 2651KLF
  • ONLINE EXCLUSIVE PRICE
    2015.00 NOK
    1669.62 NOK / Pack of 1
    Save 345.38 NOK
    17% Off
Estimated Shipment: 05-04-2027
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
  • Offer maximum flexibility for cleaning in critical environments
  • Interchangeable frames for Contec Mopping Systems
  • Autoclavable
  • Capped end and quick-connect feature for easy attachment
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mop Head Frame
Head Type Flat
Color Gray
Length (English) 7 in.
Sterility Non-sterile
Length (Metric) 18 cm
Autoclavable Autoclavable
Disposable Reusable
For Use With (Equipment) Regular VertiKlean MAX Head Frame
For Use With (Application) Designed for Use in Pharmaceutical and Medical Device Manufacturing Facilities, or Other Similar Critical or Controlled Environments.
Color-coded No
Material Stainless Steel