Learn More
Contec™ Amplitude™ Zeta™ Cleanroom Wipes
GREENER_CHOICE
Product Code. p-3138790 Shop All Contec Products
Change view
Click to view available options
Dimensions (L x W):
11 x 12 in. (27.9 x 30.5 cm)
8 x 9 in. (20.3 x 23 cm)
Unit Size:
Pack of 1500
Pack of 1800
Product Code. Dimensions (L x W) unitSize
17225176 8 x 9 in. (20.3 x 23 cm) Pack of 1800
17867283 11 x 12 in. (27.9 x 30.5 cm) Pack of 1500
2 options
Product Code. 17225176
Brand: Contec™ AMZE0001
Only null left
Add to basket
Add to basket
Prepare for acid spills. The Contec™ Amplitude™ Zeta™ Specialty Cleanroom Wipes are excellent for use with acid spills or for use as a tray liner. Wipes are lot-numbered for traceability.
  • Cellulose/polypropylene nonwoven wipes
  • Thermally-laminated, quilted wipes feature polypropylene outer layers and a highly sorptive cellulose center
  • Quilted pattern allows for segregation by appearance
  • Excellent for acid spills 
  • Ideal for tray liners
  • Lot-numbered for traceability
  • Inner core of cellulose thermally sealed between two layers of meltblown polypropylene is sorptive
  • Strong resistance to acids
  • Three-ply quilted pattern allows for segregation by appearance
Order Info
Basis weight, 103g/m2.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specialty Cleanroom Wipe
Length (English) 8 in.
Length (Metric) 20.3 cm
Width (English) 9 in.
Width (Metric) 23 cm
Material Polypropylene
Certifications/Compliance ISO Class 6-8
Dimensions (L x W) 8 x 9 in. (20.3 x 23 cm)
For Use With (Application) Cleanroom