Description
Contactin-5 Polyclonal antibody specifically detects Contactin-5 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Contactin-5 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | PE |
| Dilution | Immunocytochemistry/ Immunofluorescence |
| Formulation | PBS |
| Gene Alias | contactin 5, contactin-5, hNB-2, HNB-2s, MGC163491, NB2, NB-2, neural adhesion molecule, Neural recognition molecule NB-2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human Contactin-5 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |