Learn More
Liofilchem™ Contact Slide 2
Product Code. 11964369
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Pack
Unit Size:
Pack of 20
Product Code. Quantity unitSize
11964369 20 Pack Pack of 20
1 options
Product Code. 11964369
Brand: Liofilchem™ 525272
Rose Bengal CAF Agar and Neutralizing / PCA and TTC and Neutralizing Flex Dip-slide with a selective medium for detection of yeasts and moulds and a non selective medium for total bacterial count.
  • Contact Slide 2 is a ready-to-use device with two different media coated onto a plastic support used for the microbial monitoring of surfaces and liquids even in the presence of residues of disinfectants. The selective medium allows the isolation and enumeration of yeasts and moulds . The other medium is used for enumeration of bacteria.
Specifications
Cap Type Screw
Certifications/Compliance BD DIFCO
Manufacturing Origin Italy
Media Type Agar
Product Type Agar
Sterility Non-sterile
Target Organism Class Eukaryotic Microorganism, Fungi, Gram-positive Organisms, Fungi
For Use With (Equipment) Incubator
Incubation Conditions 10 and 25°C
Quantity 20 Pack
Species Bacteria, Yeast, Fungus