Learn More
Tork Constant Air Freshener Refill A3
GREENER_CHOICE
Product Code. p-200113531 Shop All Tork Products
Click to view available options
Scent:
Breeze
Floral
Lemongrass
Unit Size:
Pack of 6
Product Code. 30243746
Brand: Tork 257011
Tork Constant Air Freshener help to keep washrooms constantly fresh with odor neutralizer.
  • For a fresh washroom at all times, this scent contains an odour neutraliser to eliminate smells, not just mask them.
  • This propellant-free system uses only pure perfume oil, isn't flammable, and is not pressurised.
  • Use with Tork Constant Air Freshener Dispensers for long-lasting, constant scent dispersal.
Refill packs available:
  • Blossom scent with delicate floral notes for an inviting feel.
  • Breeze scent with ylang-ylang, floral and woody notes for a fresh feel.
  • Odour neutraliser scent with a very subtle, unobtrusive lemongrass smell.
  • Variety pack includes 3 each of Blossom and Breeze.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Air Freshener Refill
Alternate Part Number 7322541889337
Color-coded No
Form Spray
Scent Floral
Quantity 6/Pk.
Volume (Metric) 32 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.