missing translation for 'onlineSavingsMsg'
Learn More

Connexin-26 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16896305 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16896305 150 μL 150µL
16806315 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16896305

Brand: Proteintech 510372AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variables (EKV) is an autosomal domit genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.

Specifications

Antigen Connexin-26
Applications Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GJB2
Gene Accession No. P21994, P29033, Q00977
Gene Alias Connexin 26, Connexin-26, CX26, DFNA3, DFNA3A, DFNB1, DFNB1A, Gap junction beta 2 protein, GJB2, HID, KID, NSRD1, PPK
Gene Symbols Gjb2
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14619, 2706, 394266
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.