missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin-26 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51037-2-AP-20UL
This item is not returnable.
View return policy
Description
Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variables (EKV) is an autosomal domit genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.Specifications
| Connexin-26 | |
| Polyclonal | |
| Unconjugated | |
| GJB2 | |
| Connexin 26, Connexin-26, CX26, DFNA3, DFNA3A, DFNB1, DFNB1A, Gap junction beta 2 protein, GJB2, HID, KID, NSRD1, PPK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14619, 2706, 394266 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P21994, P29033, Q00977 | |
| Gjb2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction