Learn More
SCAT™ Conical Connector
Product Code. 10568710 Shop All SCAT Europe Products
Change view
Click to view available options
Diameter (Metric):
3 to 5 mm
4 to 8 mm
4 to 8 mm, 12 to 16 mm
4 to 8 mm, 8 to 12 mm
5 to 16 mm, 7.5 to 16 mm
7 to 10 mm
8 to 12 mm, 12 to 16 mm
Unit Size:
Each
Product Code. Diameter (Metric) unitSize
10568710 5 to 16 mm, 7.5 to 16 mm Each
10292800 4 to 8 mm Each
10011503 4 to 8 mm, 12 to 16 mm Each
10120380 8 to 12 mm, 12 to 16 mm Each
10317779 7 to 10 mm Each
10327779 3 to 5 mm Each
10820401 4 to 8 mm, 8 to 12 mm Each
7 options
Product Code. 10568710
Brand: SCAT™ 107826
Only null left
Add to basket
Add to basket
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
SCAT™ connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Capillary connectors
Specifications
Diameter (Metric) 5 to 16 mm, 7.5 to 16 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With Tubing
For Use With (Application) HPLC/UHPLC Systems
Color Transparent
Product Type Conical Connector