Description
SCAT™ connectors are available in the following versions:
- Conical connectors
- Y-connectors
- Tube connectors
- Capillary connectors
Specifications
Specifications
| Diameter (Metric) | 4 to 8 mm, 12 to 16 mm |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | Tubing |
| For Use With (Application) | HPLC/UHPLC Systems |
| Color | White |
| Product Type | Conical Connector |