Learn More
Mettler Toledo™ Connector Syringe/Filling Tube
Product Code. 15838597
Change view
Click to view available options
For Use With (Equipment):
Densito and DensitoPro Handheld Density Meters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15838597 Densito and DensitoPro Handheld Density Meters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15838597 Supplier Mettler Toledo™ Supplier No. 30330853
  • ONLINE EXCLUSIVE PRICE
    413.00 NOK
    352.16 NOK / Pack of 1
    Save 60.84 NOK
    14% Off
View all promotions
In Stock
Mettler Toledo Connector Syringe/Filling Tube are ideal for use with viscous samples
  • When working with viscous samples, this connector acts as an adapter between the Density2Go handheld meter and the syringe, making your work easier
TRUSTED_SUSTAINABILITY
Specifications
Description Connector Syringe/Filling Tube, Densito
Languages Chinese, English (GB), English, French, German, Italian, Korean, Polish, Brazilian Portuguese, Russian, Spanish
For Use With (Equipment) Densito and DensitoPro Handheld Density Meters
For Use With (Application) Use with Densito and DensitoPro Handheld Density Meters