Learn More
WTW™ Connection Cable with DIN Plug
For use with WTW™ pH/ORP electrodes with plug head.
Brand: WTW™ 108110
Additional Details : Weight : 0.04860kg
Description
- 1m long
- Supplied individually
Specifications
1 m Length | |
Connection Cable | |
DIN plug, socket S7 | |
1 m |