Learn More
BRAND™ Conical Joint Stopper
Product Code. 10653012 Shop All Brand Products
Change view
Click to view available options
Standard Taper Joint:
NS 10/19
NS 34/35
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
10653012 NS 10/19 Each
10578211 NS 34/35 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10653012 Supplier BRAND™ Supplier No. 144610
  • ONLINE EXCLUSIVE PRICE
    63.00 NOK
    57.30 NOK / Pack of 1
    Save 5.70 NOK
    9% Off
Estimated Shipment: 17-12-2026
Conical Joint Stopper features a hollow design and hexagonal grip.
  • Borosilicate Glass 3.3
  • DIN 12252
  • Hollow with hexagonal grip
TRUSTED_SUSTAINABILITY
Specifications
Description Hollow with hexagonal grip
Standard Taper Joint NS 10/19
Material Borosilicate Glass
Shape Conical