Learn More
Thermo Scientific Chemicals Al-23 Crucible, Conical;Ht (mm), 45;Top OD (mm), 35;Top ID (mm), 31;Base OD (mm), 18;Volume (ml), 23
Product Code. 15413535
Change view
Click to view available options
Capacity (Metric):
4 mL
5 mL
8 mL
9 mL
23 mL
40 mL
49 mL
75 mL
125 mL
160 mL
Diameter (Metric) Outer:
17mm
21mm
22mm
26mm
31mm
40mm
45mm
53mm
67mm
80mm
Diameter (Metric) Inner:
20mm
24mm
25mm
30mm
40mm
45mm
55mm
60mm
65mm
90mm
Height (Metric):
20mm (Top), 11mm (Base)
25mm (Top), 13mm (Base)
30mm (Top), 15mm (Base)
35mm (Top), 18mm (Base)
45mm (Top), 22mm (Base)
50mm (Top), 25mm (Base)
60mm (Top), 30mm (Base)
73mm (Top), 35mm (Base)
88mm (Top), 44mm (Base)
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Outer Diameter (Metric) Inner Height (Metric) unitSize
15413535 49 mL 40mm 55mm 45mm (Top), 22mm (Base) Each
15483525 9 mL 21mm 30mm 25mm (Top), 13mm (Base) Each
15473525 4 mL 17mm 25mm 20mm (Top), 11mm (Base) Each
15423535 75 mL 53mm 65mm 60mm (Top), 30mm (Base) Each
15403535 23 mL 31mm 45mm 35mm (Top), 18mm (Base) Each
15433535 160 mL 67mm 90mm 73mm (Top), 35mm (Base) Each
15453535 8 mL 26mm 24mm 30mm (Top), 15mm (Base) Each
15443535 5 mL 22mm 20mm 25mm (Top), 13mm (Base) Each
15493535 125 mL 80mm 60mm 88mm (Top), 44mm (Base) Each
15473535 40 mL 45mm 40mm 50mm (Top), 25mm (Base) Each
10 options
Product Code. 15413535
Brand: Thermo Scientific Chemicals 033013.KF
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 55mm
Material Al-23
Quantity 1 Piece
Capacity (Metric) 49 mL
Diameter (Metric) Outer 40mm
Height (Metric) 45mm (Top), 22mm (Base)
Product Type Conical Al-23 Crucibles
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.